Product Details

SNP ID
rs11555424
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:201957519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTTTTTGTTATAGGGAGTAAA[C/T]CACAGACTACGAGGGAGTTTGACAG
Phenotype
MIM: 605057
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TIMM17A PubMed Links

Gene Details

Gene
TIMM17A
Gene Name
translocase of inner mitochondrial membrane 17 homolog A (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006335.2 171 Silent Mutation AAC,AAT N45N NP_006326.1

View Full Product Details