Product Details

SNP ID
rs28362414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10441287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAAGCAAGCGTTTCTTCAGTCAC[C/T]GTTTTGGTAATAAGGAAACCACAAG
Phenotype
MIM: 165640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929715 PubMed Links

Gene Details

Gene
LOC101929715
Gene Name
uncharacterized LOC101929715
There are no transcripts associated with this gene.

Gene
ODC1
Gene Name
ornithine decarboxylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287188.1 Intron NP_001274117.1
NM_001287189.1 Intron NP_001274118.1
NM_001287190.1 Intron NP_001274119.1
NM_002539.2 Intron NP_002530.1
Gene
SNORA80B
Gene Name
small nucleolar RNA, H/ACA box 80B
There are no transcripts associated with this gene.

View Full Product Details