Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004610.3 | Intron | NP_004601.3 | |||
XM_011536082.2 | Intron | XP_011534384.1 | |||
XM_011536084.2 | Intron | XP_011534386.1 | |||
XM_011536085.2 | Intron | XP_011534387.1 | |||
XM_011536088.2 | Intron | XP_011534390.1 | |||
XM_011536090.2 | Intron | XP_011534392.1 | |||
XM_017011228.1 | Intron | XP_016866717.1 | |||
XM_017011229.1 | Intron | XP_016866718.1 | |||
XM_017011230.1 | Intron | XP_016866719.1 | |||
XM_017011231.1 | Intron | XP_016866720.1 |