Product Details

SNP ID
rs41563617
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29943022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCGGCTACTACAACCAGAGCGA[A/G]GCCGGTGAGTGACCCCGGCCGGGGG
Phenotype
MIM: 142800
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HLA-A PubMed Links

Gene Details

Gene
HLA-A
Gene Name
major histocompatibility complex, class I, A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242758.1 423 Silent Mutation GAA,GAG E113E NP_001229687.1
NM_002116.7 423 Silent Mutation GAA,GAG E113E NP_002107.3

View Full Product Details