Product Details

SNP ID
rs35852819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31804615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTATCATATAACTGGCCTATGC[A/T]AGGTATTAGATACACTACATGGTTT
Phenotype
MIM: 140559 MIM: 607282 MIM: 192150
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HSPA1L PubMed Links

Gene Details

Gene
HSPA1L
Gene Name
heat shock protein family A (Hsp70) member 1 like
There are no transcripts associated with this gene.

Gene
LSM2
Gene Name
LSM2 homolog, U6 small nuclear RNA and mRNA degradation associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021177.4 Intron NP_067000.1
Gene
VARS
Gene Name
valyl-tRNA synthetase
There are no transcripts associated with this gene.

View Full Product Details