Product Details

SNP ID
rs17883943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:53498092 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTATAGAGTTGTGTTTAAACTTAA[C/T]TCCTGCATCAGCAAATTTAAACAAA
Phenotype
MIM: 606857
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCLC PubMed Links

Gene Details

Gene
GCLC
Gene Name
glutamate-cysteine ligase catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197115.1 2687 UTR 3 NP_001184044.1
NM_001498.3 2687 UTR 3 NP_001489.1
XM_017010749.1 2687 UTR 3 XP_016866238.1

View Full Product Details