Product Details

SNP ID
rs35913626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2242571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCGGGACAAAGTACACGGGCCTG[A/G]TGTGAAACTAAGCCACAAACCCATT
Phenotype
MIM: 602686 MIM: 606906 MIM: 600312 MIM: 614905
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAD1L1 PubMed Links

Gene Details

Gene
MAD1L1
Gene Name
MAD1 mitotic arrest deficient like 1
There are no transcripts associated with this gene.

Gene
MRM2
Gene Name
mitochondrial rRNA methyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013393.1 Intron NP_037525.1
Gene
NUDT1
Gene Name
nudix hydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002452.3 Intron NP_002443.3
NM_198948.1 Intron NP_945186.1
NM_198949.1 Intron NP_945187.1
NM_198950.1 Intron NP_945188.1
NM_198952.1 Intron NP_945190.1
NM_198953.1 Intron NP_945191.1
NM_198954.1 Intron NP_945192.1
Gene
SNX8
Gene Name
sorting nexin 8
There are no transcripts associated with this gene.

View Full Product Details