Product Details
- SNP ID
-
rs28709565
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:35233097 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTCAAAGTATATCTATAGAACACT[G/T]TAAACTGTTAACACGTGAGGAATTT
- Phenotype
-
MIM: 606061
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TBX20
PubMed Links
Gene Details
- Gene
- TBX20
- Gene Name
- T-box 20
View Full Product Details