Product Details
- SNP ID
-
rs17885558
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:95359938 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATCAGTTTACTTTTACAAAATAT[A/G]TAGACTTTTTTTTTTTTTTTTTACT
- Phenotype
-
MIM: 602720
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PON3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs553882161] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PON3
- Gene Name
- paraoxonase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000940.2 |
1125 |
UTR 3 |
|
|
NP_000931.1 |
View Full Product Details