Product Details

SNP ID
rs28659815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99031121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGGAGAAATTCTTTCTCTGTAC[G/T]GGGAATGCCTCTTTGGCAGCCCCTG
Phenotype
MIM: 605568
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927550 PubMed Links

Gene Details

Gene
LOC101927550
Gene Name
uncharacterized LOC101927550
There are no transcripts associated with this gene.

Gene
SMURF1
Gene Name
SMAD specific E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199847.1 Intron NP_001186776.1
NM_020429.2 Intron NP_065162.1
NM_181349.2 Intron NP_851994.1
XM_017012457.1 Intron XP_016867946.1

View Full Product Details