Product Details
- SNP ID
-
rs28495024
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:99419060 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCCCCAAACAGTAGCAGAGCCAC[C/G]TCTGCTCATCGCAGGGTCTGTCATG
- Phenotype
-
MIM: 603477
MIM: 614774
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP5J2-PTCD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117194181] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP5J2-PTCD1
- Gene Name
- ATP5J2-PTCD1 readthrough
- Gene
- BUD31
- Gene Name
- BUD31 homolog
- Gene
- PTCD1
- Gene Name
- pentatricopeptide repeat domain 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015545.3 |
3184 |
UTR 3 |
|
|
NP_056360.2 |
View Full Product Details