Product Details

SNP ID
rs28365075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99649084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGTAACATTCTGATCTCAGTTC[A/T]GTGAGGGGAAAGCAAATTGCCATGG
Phenotype
MIM: 605325
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP3A5 PubMed Links

Gene Details

Gene
CYP3A5
Gene Name
cytochrome P450 family 3 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000777.4 Intron NP_000768.1
NM_001190484.2 Intron NP_001177413.1
NM_001291829.1 Intron NP_001278758.1
NM_001291830.1 Intron NP_001278759.1
XM_006715859.2 Intron XP_006715922.1
XM_011515843.1 Intron XP_011514145.1
XM_017011781.1 Intron XP_016867270.1
XM_017011782.1 Intron XP_016867271.1
Gene
ZSCAN25
Gene Name
zinc finger and SCAN domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145115.2 Intron NP_660090.2
XM_005250194.2 Intron XP_005250251.1
XM_011515905.2 Intron XP_011514207.1
XM_011515906.2 Intron XP_011514208.1
XM_011515907.2 Intron XP_011514209.1
XM_011515908.2 Intron XP_011514210.1
XM_011515909.2 Intron XP_011514211.1
XM_011515910.2 Intron XP_011514212.1
XM_017011824.1 Intron XP_016867313.1
XM_017011825.1 Intron XP_016867314.1
XM_017011826.1 Intron XP_016867315.1
XM_017011827.1 Intron XP_016867316.1

View Full Product Details