Product Details

SNP ID
rs28718326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:113082694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAATAAATGATCTTCCATGCAC[G/T]TTTAGATGCATGACCATGACCAGTT
Phenotype
MIM: 605188
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR85 PubMed Links

Gene Details

Gene
GPR85
Gene Name
G protein-coupled receptor 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146265.1 2717 UTR 3 NP_001139737.1
NM_001146266.1 2717 UTR 3 NP_001139738.1
NM_001146267.1 2717 UTR 3 NP_001139739.1
NM_018970.6 2717 UTR 3 NP_061843.3
XM_005250451.2 2717 UTR 3 XP_005250508.1
XM_017012352.1 2717 UTR 3 XP_016867841.1

View Full Product Details