Product Details

SNP ID
rs28371048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:135364104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACAAAAAAATGAAAGATAAAAA[A/T]AAATAAAAAGCTACGTTAGAAACAT
Phenotype
MIM: 604911
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CNOT4 PubMed Links

Gene Details

Gene
CNOT4
Gene Name
CCR4-NOT transcription complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008225.2 Intron NP_001008226.1
NM_001190847.1 Intron NP_001177776.1
NM_001190848.1 Intron NP_001177777.1
NM_001190849.1 Intron NP_001177778.1
NM_001190850.1 Intron NP_001177779.1
NM_013316.3 Intron NP_037448.2
XM_017012235.1 Intron XP_016867724.1

View Full Product Details