Product Details

SNP ID
rs28526625
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:139135499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTAAATAATTCTAACATGCTTAC[A/G]GTTGAAAATATAGCCCCAGAAAATA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TTC26 PubMed Links

Gene Details

Gene
TTC26
Gene Name
tetratricopeptide repeat domain 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144920.2 Intron NP_001138392.1
NM_001144923.2 Intron NP_001138395.1
NM_001287512.1 Intron NP_001274441.1
NM_001287513.1 Intron NP_001274442.1
NM_001318333.1 Intron NP_001305262.1
NM_001321740.1 Intron NP_001308669.1
NM_001321741.1 Intron NP_001308670.1
NM_001321742.1 Intron NP_001308671.1
NM_024926.3 Intron NP_079202.2

View Full Product Details