Product Details

SNP ID
rs28451965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86468805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCAAGACCTACAGGCAGAGCTC[C/T]TGATTTATCTACCCAGTAGCAGTCA
Phenotype
MIM: 611871 MIM: 602307
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RMDN1 PubMed Links

Gene Details

Gene
RMDN1
Gene Name
regulator of microtubule dynamics 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286707.1 Intron NP_001273636.1
NM_001286719.1 Intron NP_001273648.1
NM_001317807.1 Intron NP_001304736.1
NM_016033.2 Intron NP_057117.2
XM_005250944.3 Intron XP_005251001.1
XM_005250947.3 Intron XP_005251004.1
XM_005250948.3 Intron XP_005251005.1
XM_006716573.3 Intron XP_006716636.1
XM_011517090.2 Intron XP_011515392.1
XM_011517091.2 Intron XP_011515393.1
XM_011517092.2 Intron XP_011515394.1
XM_011517093.2 Intron XP_011515395.1
XM_011517094.2 Intron XP_011515396.1
XM_011517095.2 Intron XP_011515397.1
XM_011517096.2 Intron XP_011515398.1
XM_011517097.2 Intron XP_011515399.1
XM_011517098.2 Intron XP_011515400.1
XM_017013518.1 Intron XP_016869007.1
XM_017013519.1 Intron XP_016869008.1
XM_017013520.1 Intron XP_016869009.1
XM_017013521.1 Intron XP_016869010.1
XM_017013522.1 Intron XP_016869011.1
XM_017013523.1 Intron XP_016869012.1
XM_017013524.1 Intron XP_016869013.1
XM_017013525.1 Intron XP_016869014.1
XM_017013526.1 Intron XP_016869015.1
XM_017013527.1 Intron XP_016869016.1
XM_017013528.1 Intron XP_016869017.1
Gene
WWP1
Gene Name
WW domain containing E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007013.3 Intron NP_008944.1
XM_005250760.3 Intron XP_005250817.1
XM_005250761.3 Intron XP_005250818.1
XM_011516802.2 Intron XP_011515104.1
XM_017012993.1 Intron XP_016868482.1
XM_017012994.1 Intron XP_016868483.1
XM_017012995.1 Intron XP_016868484.1
XM_017012996.1 Intron XP_016868485.1
XM_017012997.1 Intron XP_016868486.1

View Full Product Details