Product Details

SNP ID
rs28362550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:13107462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCAATAGAATCCAGGATTTTCT[A/G]TCCTGTAAATAAGACCTACAGAAAG
Phenotype
MIM: 603785
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MPDZ PubMed Links

Gene Details

Gene
MPDZ
Gene Name
multiple PDZ domain crumbs cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261406.1 Intron NP_001248335.1
NM_001261407.1 Intron NP_001248336.1
NM_003829.4 Intron NP_003820.2
XM_005251622.4 Intron XP_005251679.1
XM_005251623.4 Intron XP_005251680.1
XM_006716885.3 Intron XP_006716948.1
XM_006716886.3 Intron XP_006716949.1
XM_006716887.3 Intron XP_006716950.1
XM_006716888.3 Intron XP_006716951.1
XM_006716889.3 Intron XP_006716952.1
XM_006716891.3 Intron XP_006716954.1
XM_017015252.1 Intron XP_016870741.1
XM_017015253.1 Intron XP_016870742.1
XM_017015254.1 Intron XP_016870743.1
XM_017015255.1 Intron XP_016870744.1
XM_017015256.1 Intron XP_016870745.1
XM_017015257.1 Intron XP_016870746.1
XM_017015258.1 Intron XP_016870747.1
XM_017015259.1 Intron XP_016870748.1

View Full Product Details