Product Details

SNP ID
rs34754082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:43882121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGACAAATACCTGTATCCATCC[G/T]AAATCAGATTTTCTGAAGGTGGGCC
Phenotype
MIM: 610603
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALKBH3 PubMed Links

Gene Details

Gene
ALKBH3
Gene Name
alkB homolog 3, alpha-ketoglutaratedependent dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139178.3 Intron NP_631917.1

View Full Product Details