Product Details

SNP ID
hCV59892925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10881039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTCACGGCATTTGAAACACTGT[C/T]ATCTTCTTATTCACTTCCTGAAAAA
Phenotype
MIM: 168730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRH1 PubMed Links

Gene Details

Gene
PRH1
Gene Name
proline rich protein HaeIII subfamily 1
There are no transcripts associated with this gene.

Gene
PRH1-PRR4
Gene Name
PRH1-PRR4 readthrough
There are no transcripts associated with this gene.

View Full Product Details