Product Details

SNP ID
rs1669413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:11186175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCCACAACATAAAAAATATCC[A/C]ATTGGCCACTTGTAAGGAAAAAAAA
Phenotype
MIM: 613966
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TAS2R42 PubMed Links

Gene Details

Gene
TAS2R42
Gene Name
taste 2 receptor member 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181429.2 763 Missense Mutation GGG,TGG G255W NP_852094.2

View Full Product Details