Product Details
- SNP ID
-
rs28729376
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:48824576 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTCTAAACTATGTGAATGTTGAT[C/G]TCAGGTGTGCCATTCATTCTGTGAG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SHC4
PubMed Links
Gene Details
- Gene
- SHC4
- Gene Name
- SHC adaptor protein 4
View Full Product Details