Product Details

SNP ID
rs28504405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73994894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCCACCATGCCTCCCCCCGAG[A/C]CCCCCTCTGAAGGCCGCCAGCCCAG
Phenotype
MIM: 102578 MIM: 608326
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PML PubMed Links

Gene Details

Gene
PML
Gene Name
promyelocytic leukemia
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002675.3 222 Missense Mutation ACC,CCC T28P NP_002666.1
NM_033238.2 222 Missense Mutation ACC,CCC T28P NP_150241.2
NM_033239.2 222 Missense Mutation ACC,CCC T28P NP_150242.1
NM_033240.2 222 Missense Mutation ACC,CCC T28P NP_150243.2
NM_033244.3 222 Missense Mutation ACC,CCC T28P NP_150247.2
NM_033246.2 222 Missense Mutation ACC,CCC T28P NP_150249.1
NM_033247.2 222 Missense Mutation ACC,CCC T28P NP_150250.2
NM_033249.2 222 Missense Mutation ACC,CCC T28P NP_150252.1
NM_033250.2 222 Missense Mutation ACC,CCC T28P NP_150253.2
Gene
STOML1
Gene Name
stomatin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256672.1 222 Intron NP_001243601.1
NM_001256673.1 222 Intron NP_001243602.1
NM_001256674.1 222 Intron NP_001243603.1
NM_001256675.1 222 Intron NP_001243604.1
NM_001256676.1 222 Intron NP_001243605.1
NM_001256677.1 222 Intron NP_001243606.1
NM_001324226.1 222 Intron NP_001311155.1
NM_001324227.1 222 Intron NP_001311156.1
NM_001324228.1 222 Intron NP_001311157.1
NM_001324229.1 222 Intron NP_001311158.1
NM_001324230.1 222 Intron NP_001311159.1
NM_004809.4 222 Intron NP_004800.2
XM_005254792.4 222 Intron XP_005254849.1
XM_006720773.3 222 Intron XP_006720836.1

View Full Product Details