Product Details

SNP ID
rs28412621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73998109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTGTCTGCACACGCTGTGCTCA[C/G]GATGCCTGGAGGCGTCGGGCATGCA
Phenotype
MIM: 102578 MIM: 608326
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PML PubMed Links

Gene Details

Gene
PML
Gene Name
promyelocytic leukemia
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002675.3 375 Missense Mutation CGA,GGA R79G NP_002666.1
NM_033238.2 375 Missense Mutation CGA,GGA R79G NP_150241.2
NM_033239.2 375 Missense Mutation CGA,GGA R79G NP_150242.1
NM_033240.2 375 Missense Mutation CGA,GGA R79G NP_150243.2
NM_033244.3 375 Missense Mutation CGA,GGA R79G NP_150247.2
NM_033246.2 375 Missense Mutation CGA,GGA R79G NP_150249.1
NM_033247.2 375 Missense Mutation CGA,GGA R79G NP_150250.2
NM_033249.2 375 Missense Mutation CGA,GGA R79G NP_150252.1
NM_033250.2 375 Missense Mutation CGA,GGA R79G NP_150253.2
Gene
STOML1
Gene Name
stomatin like 1
There are no transcripts associated with this gene.

View Full Product Details