Product Details

SNP ID
rs28655943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:38459954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGTTGGATGAGGGGCAAAGACA[A/G]TAGGTTGGGTAAATGATGGGAAAAG
Phenotype
MIM: 616142
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM98B PubMed Links

Gene Details

Gene
FAM98B
Gene Name
family with sequence similarity 98 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173611.3 Intron NP_775882.2

View Full Product Details