Product Details

SNP ID
rs35168952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30477149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGCATCCTTCTGTGTGAGAGA[C/T]AGAAGACTGAGTGAAATATTCATCC
Phenotype
MIM: 153370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGAL PubMed Links

Gene Details

Gene
ITGAL
Gene Name
integrin subunit alpha L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114380.1 Intron NP_001107852.1
NM_002209.2 Intron NP_002200.2
XM_005255313.1 Intron XP_005255370.1
XM_006721044.1 Intron XP_006721107.1
XM_011545849.1 Intron XP_011544151.1

View Full Product Details