Product Details

SNP ID
rs17883103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70461526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCCTACTTGCTGAGAGTGCCC[G/T]GGAGGAAGATGGCGGGCAGGTAGGG
Phenotype
MIM: 608675
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FUK PubMed Links

Gene Details

Gene
FUK
Gene Name
fucokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145059.2 Intron NP_659496.2
XM_006721162.1 Intron XP_006721225.1
XM_011522928.1 Intron XP_011521230.1
XM_011522929.1 Intron XP_011521231.1
XM_011522930.2 Intron XP_011521232.1
XM_011522931.2 Intron XP_011521233.1
XM_017023012.1 Intron XP_016878501.1
XM_017023013.1 Intron XP_016878502.1
XM_017023014.1 Intron XP_016878503.1

View Full Product Details