Product Details

SNP ID
rs28479510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74874945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTCTTCTGAGCCACTGCACCC[A/G]GCTGCTGAGTACTGTATTTTCTCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR59 PubMed Links

Gene Details

Gene
WDR59
Gene Name
WD repeat domain 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324171.1 Intron NP_001311100.1
NM_001324172.1 Intron NP_001311101.1
NM_030581.3 Intron NP_085058.3
XM_005256146.3 Intron XP_005256203.1
XM_011523331.2 Intron XP_011521633.1
XM_011523332.2 Intron XP_011521634.1
XM_017023667.1 Intron XP_016879156.1
XM_017023668.1 Intron XP_016879157.1
XM_017023669.1 Intron XP_016879158.1
XM_017023670.1 Intron XP_016879159.1

View Full Product Details