Product Details
- SNP ID
-
rs28778946
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:81103600 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGCTGGGGAATGGATAAGCACAT[C/G]GTTGTTTTACCCACACAATGAAATA
- Phenotype
-
MIM: 238330
MIM: 607894
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GCSH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79546329] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GCSH
- Gene Name
- glycine cleavage system protein H
There are no transcripts associated with this gene.
- Gene
- PKD1L2
- Gene Name
- polycystin 1 like 2 (gene/pseudogene)
View Full Product Details