Product Details

SNP ID
rs28362626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74431533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCGCGCTCCGAGTTGGGCAGAAC[C/T]TGGAACGGTCACAGGAAACCCGGGC
Phenotype
MIM: 605949
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPRC5C PubMed Links

Gene Details

Gene
GPRC5C
Gene Name
G protein-coupled receptor class C group 5 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018653.3 6 Intron NP_061123.3
NM_022036.2 6 UTR 5 NP_071319.2
XM_005257512.1 6 Intron XP_005257569.1
XM_005257514.1 6 UTR 5 XP_005257571.1
XM_017024855.1 6 Intron XP_016880344.1
XM_017024856.1 6 Intron XP_016880345.1

View Full Product Details