Product Details

SNP ID
rs3210169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81845595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACTGGCAGGAAAGAACTTGAGT[C/G]TGGGGAAGCTGTGCACTTTGACGGC
Phenotype
MIM: 176790
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
P4HB PubMed Links

Gene Details

Gene
P4HB
Gene Name
prolyl 4-hydroxylase subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000918.3 1522 Missense Mutation ACA,AGA T442R NP_000909.2

View Full Product Details