Product Details

SNP ID
rs1128505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42609765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGAGGGAAATCATCACCCTACAG[C/T]TGGGCCAGTGCGGCAATCAGAGTGA
Phenotype
MIM: 616498 MIM: 191135
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM134C PubMed Links

Gene Details

Gene
FAM134C
Gene Name
family with sequence similarity 134 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178126.3 426 Intron NP_835227.1
XM_011524440.2 426 Intron XP_011522742.1
Gene
TUBG1
Gene Name
tubulin gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001070.4 426 Silent Mutation CTG,TTG L10L NP_001061.2

View Full Product Details