Product Details

SNP ID
rs33954211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42851357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCGTGGGAAAATGAACCAGAAGA[C/G]AATCCTCGTGCTCCTCATTCTGGCC
Phenotype
MIM: 602268 MIM: 603735 MIM: 605129
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AOC2 PubMed Links

Gene Details

Gene
AOC2
Gene Name
amine oxidase, copper containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001158.3 235 Intron NP_001149.2
NM_009590.2 235 Intron NP_033720.2
Gene
AOC3
Gene Name
amine oxidase, copper containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277731.1 235 Missense Mutation ACA,AGA T5R NP_001264660.1
NM_001277732.1 235 Intron NP_001264661.1
NM_003734.3 235 Missense Mutation ACA,AGA T5R NP_003725.1
XM_011525419.1 235 Missense Mutation ACA,AGA T5R XP_011523721.1
XM_011525420.2 235 Missense Mutation ACA,AGA T5R XP_011523722.1
Gene
PSME3
Gene Name
proteasome activator subunit 3
There are no transcripts associated with this gene.

View Full Product Details