Product Details

SNP ID
rs34361243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45076890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATATAAAGAAAATGAAATTTGC[C/T]CTCTGATTTTAGTTTCGGTCTTGGA
Phenotype
MIM: 160993
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NMT1 PubMed Links

Gene Details

Gene
NMT1
Gene Name
N-myristoyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021079.4 Intron NP_066565.1
XM_011524858.2 Intron XP_011523160.1

View Full Product Details