Product Details

SNP ID
hCV60558273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48070086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAATCTATAGTTTTATTAAGACA[A/G]AAACTGACAATGTAGTATGAAGTTT
Phenotype
MIM: 604511 MIM: 163260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBX1 PubMed Links

Gene Details

Gene
CBX1
Gene Name
chromobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127228.1 2388 Intron NP_001120700.1
NM_006807.4 2388 UTR 3 NP_006798.1
Gene
NFE2L1
Gene Name
nuclear factor, erythroid 2 like 1
There are no transcripts associated with this gene.

View Full Product Details