Product Details

SNP ID
rs28370320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48721715 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTATAATCACAGTTAACTTTCAC[C/T]CAGTTTCCTTTTTTAAAAAAATTTT
Phenotype
MIM: 604607 MIM: 609819 MIM: 610787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXB13 PubMed Links

Gene Details

Gene
HOXB13
Gene Name
homeobox B13
There are no transcripts associated with this gene.

Gene
MIR3185
Gene Name
microRNA 3185
There are no transcripts associated with this gene.

Gene
PRAC1
Gene Name
prostate cancer susceptibility candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032391.2 Intron NP_115767.1
Gene
PRAC2
Gene Name
prostate cancer susceptibility candidate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282275.1 Intron NP_001269204.1
NM_001282276.1 Intron NP_001269205.1
XM_006721865.3 Intron XP_006721928.1
XM_011524747.2 Intron XP_011523049.1
XM_011524748.1 Intron XP_011523050.1

View Full Product Details