Product Details

SNP ID
rs28753938
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:79978272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATGAACCTTTACAAATAGTTACA[C/T]GCATCTCACAACTGCTCTAAAATAT
Phenotype
MIM: 611595
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HSBP1L1 PubMed Links

Gene Details

Gene
HSBP1L1
Gene Name
heat shock factor binding protein 1 like 1
There are no transcripts associated with this gene.

Gene
TXNL4A
Gene Name
thioredoxin like 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303471.2 Intron NP_001290400.1
NM_001305557.1 Intron NP_001292486.1
NM_001305563.1 Intron NP_001292492.1
NM_001305564.1 Intron NP_001292493.1
NM_006701.4 Intron NP_006692.1

View Full Product Details