Product Details

SNP ID
rs35946201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48955122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAGTCCCCTCCGTCCTCGGAGG[A/T]TCCTGGCTCTCTGATCCCCGTGTCC
Phenotype
MIM: 600040
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BAX PubMed Links

Gene Details

Gene
BAX
Gene Name
BCL2 associated X, apoptosis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291428.1 Intron NP_001278357.1
NM_001291429.1 Intron NP_001278358.1
NM_001291430.1 Intron NP_001278359.1
NM_001291431.1 Intron NP_001278360.1
NM_004324.3 Intron NP_004315.1
NM_138761.3 Intron NP_620116.1
NM_138763.3 Intron NP_620118.1
NM_138764.4 Intron NP_620119.2
XM_017027077.1 Intron XP_016882566.1

View Full Product Details