Product Details

SNP ID
rs34124134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48955841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACATGGAGCTGCAGAGGTGTGGG[C/T]CCCTGAGGACCCAGAAGTCCAGCCA
Phenotype
MIM: 600040 MIM: 134790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAX PubMed Links

Gene Details

Gene
BAX
Gene Name
BCL2 associated X, apoptosis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291428.1 Intron NP_001278357.1
NM_001291429.1 Intron NP_001278358.1
NM_001291430.1 Intron NP_001278359.1
NM_001291431.1 Intron NP_001278360.1
NM_004324.3 Intron NP_004315.1
NM_138761.3 Intron NP_620116.1
NM_138763.3 Intron NP_620118.1
NM_138764.4 Intron NP_620119.2
XM_017027077.1 Intron XP_016882566.1
Gene
FTL
Gene Name
ferritin light chain
There are no transcripts associated with this gene.

View Full Product Details