Product Details
- SNP ID
-
rs28586965
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:49906897 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTATTTCAGAGAATTCAGCAAAGT[A/G]TAAAGACAAATACAAAAGATGCTTG
- Phenotype
-
MIM: 609742
MIM: 605815
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IL4I1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115030648] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IL4I1
- Gene Name
- interleukin 4 induced 1
- Gene
- NUP62
- Gene Name
- nucleoporin 62
View Full Product Details