Product Details

SNP ID
rs28666068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54617261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGCAAGAAGAGTGACCCCCTTG[A/T]GGGCACTCCATTGGTTTTATGGCGC
Phenotype
MIM: 604811
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LILRB1 PubMed Links

Gene Details

Gene
LILRB1
Gene Name
leukocyte immunoglobulin like receptor B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081637.2 323 Intron NP_001075106.2
NM_001081638.3 323 Intron NP_001075107.2
NM_001081639.3 323 Intron NP_001075108.2
NM_001278398.2 323 Intron NP_001265327.2
NM_001278399.2 323 Intron NP_001265328.2
NM_006669.6 323 UTR 5 NP_006660.4
XM_011526331.2 323 Intron XP_011524633.1
XM_011526332.2 323 Intron XP_011524634.1
XM_011526335.2 323 Intron XP_011524637.1
XM_011526336.2 323 Intron XP_011524638.1
XM_011526338.2 323 Intron XP_011524640.1
XM_017026182.1 323 Intron XP_016881671.1
XM_017026183.1 323 Intron XP_016881672.1
XM_017026184.1 323 Intron XP_016881673.1
XM_017026185.1 323 Intron XP_016881674.1
XM_017026186.1 323 Intron XP_016881675.1
XM_017026187.1 323 UTR 5 XP_016881676.1
XM_017026188.1 323 Intron XP_016881677.1
XM_017026189.1 323 Intron XP_016881678.1
XM_017026190.1 323 UTR 5 XP_016881679.1
XM_017026191.1 323 Intron XP_016881680.1
XM_017026192.1 323 Intron XP_016881681.1

View Full Product Details