Product Details

SNP ID
rs28508429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35295368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACTTCCCCCAGCCTTTAACCC[C/T]CTCCTCTCCCTTTCCAGCGATCACT
Phenotype
MIM: 159460
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAG PubMed Links

Gene Details

Gene
MAG
Gene Name
myelin associated glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199216.1 Intron NP_001186145.1
NM_002361.3 Intron NP_002352.1
NM_080600.2 Intron NP_542167.1

View Full Product Details