Product Details

SNP ID
rs34537352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3607824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTATTCCCAGCCTGGCCCGGCCC[G/T]CTCGTCAGCCCAGTCCGGAAGCCCC
Phenotype
MIM: 188070
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CACTIN PubMed Links

Gene Details

Gene
CACTIN
Gene Name
cactin, spliceosome C complex subunit
There are no transcripts associated with this gene.

Gene
CACTIN-AS1
Gene Name
CACTIN antisense RNA 1
There are no transcripts associated with this gene.

Gene
TBXA2R
Gene Name
thromboxane A2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001060.5 926 Intron NP_001051.1
NM_201636.2 926 Intron NP_963998.2
XM_011528214.2 926 UTR 5 XP_011526516.1

View Full Product Details