Product Details

SNP ID
rs28382791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10393554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCGCCCCACGGTTCCCCAAGTC[C/T]ATTCCTGACCTACATGAAGGGCTCC
Phenotype
MIM: 605065
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC37 PubMed Links

Gene Details

Gene
CDC37
Gene Name
cell division cycle 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007065.3 Intron NP_008996.1
XM_011527652.2 Intron XP_011525954.1
Gene
MIR1181
Gene Name
microRNA 1181
There are no transcripts associated with this gene.

View Full Product Details