Product Details

SNP ID
rs11550960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15834602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAATGGACCAGACCCTACCCGGT[C/T]ATTTATAGCACACCAACTGCCGTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UCA1 PubMed Links

Gene Details

Gene
UCA1
Gene Name
urothelial cancer associated 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details