Product Details

SNP ID
rs17878543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18059170 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGTGGGGGGTGGGATGGGAGGAC[C/T]GTCATTTTTCCCACAAAGTTTACAG
Phenotype
MIM: 601604
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL12RB1 PubMed Links

Gene Details

Gene
IL12RB1
Gene Name
interleukin 12 receptor subunit beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290023.1 2972 UTR 3 NP_001276952.1
NM_001290024.1 2972 UTR 3 NP_001276953.1
NM_005535.2 2972 UTR 3 NP_005526.1
NM_153701.2 2972 Intron NP_714912.1
XM_006722741.3 2972 UTR 3 XP_006722804.2
XM_011527966.2 2972 UTR 3 XP_011526268.1
XM_011527967.2 2972 UTR 3 XP_011526269.1
XM_011527968.2 2972 UTR 3 XP_011526270.1
XM_011527969.2 2972 UTR 3 XP_011526271.1
XM_011527970.2 2972 UTR 3 XP_011526272.1
XM_011527971.2 2972 UTR 3 XP_011526273.1
XM_011527972.2 2972 UTR 3 XP_011526274.1
XM_011527973.2 2972 UTR 3 XP_011526275.1
XM_011527974.2 2972 UTR 3 XP_011526276.1
XM_011527975.2 2972 UTR 3 XP_011526277.1
XM_011527976.2 2972 Intron XP_011526278.1
XM_011527977.2 2972 Intron XP_011526279.1
XM_017026762.1 2972 UTR 3 XP_016882251.1

View Full Product Details