Product Details
- SNP ID
-
rs28694254
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:41171213 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGACCCTCCGGAACTCCGTGCTGTT[C/T]CCCCATCTCCTCTTTCCTTCAGCAC
- Phenotype
-
MIM: 605668
MIM: 613770
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BACE2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114441610] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BACE2
- Gene Name
- beta-site APP-cleaving enzyme 2
- Gene
- MIR3197
- Gene Name
- microRNA 3197
There are no transcripts associated with this gene.
- Gene
- PLAC4
- Gene Name
- placenta specific 4
There are no transcripts associated with this gene.
View Full Product Details