Product Details

SNP ID
rs28488527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19331225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTAGCTCCGCATCGGGGGCTTGG[A/G]GGGAGGGATGAGCTTCCCCCTCCTG
Phenotype
MIM: 600237
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIRA PubMed Links

Gene Details

Gene
HIRA
Gene Name
histone cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003325.3 3489 UTR 3 NP_003316.3

View Full Product Details