Product Details
- SNP ID
-
rs10427923
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:20690650 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACGCAAGTTTCCCCTGCTGCCAC[A/G]CAGGCGAGGGGAGCCTTTGATGCTG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
POM121L4P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143362596] are located under a probe and SNP(s) [rs148349604] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- POM121L4P
- Gene Name
- POM121 transmembrane nucleoporin like 4, pseudogene
There are no transcripts associated with this gene.
View Full Product Details