Product Details

SNP ID
rs28427478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101392471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATTAAGGAATGACTTTAGTGCT[C/T]ATTTGGATGCAGTAAGTGGTTTGAT
Phenotype
MIM: 300300 MIM: 300644 MIM: 300902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTK PubMed Links

Gene Details

Gene
BTK
Gene Name
Bruton tyrosine kinase
There are no transcripts associated with this gene.

Gene
GLA
Gene Name
galactosidase alpha
There are no transcripts associated with this gene.

Gene
RPL36A
Gene Name
ribosomal protein L36a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199972.1 Intron NP_001186901.1
NM_021029.5 Intron NP_066357.2
Gene
RPL36A-HNRNPH2
Gene Name
RPL36A-HNRNPH2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199973.1 Intron NP_001186902.1
NM_001199974.1 Intron NP_001186903.1

View Full Product Details