Product Details

SNP ID
rs28664702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:68503061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGCTGGTCTTGAACTATAGTC[G/T]GGGATCTTTGGAAGATCTCAGGCTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
YIPF6 PubMed Links

Gene Details

Gene
YIPF6
Gene Name
Yip1 domain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195214.1 Intron NP_001182143.1
NM_173834.3 Intron NP_776195.2
XM_011530923.2 Intron XP_011529225.1
XM_011530924.1 Intron XP_011529226.1
XM_017029450.1 Intron XP_016884939.1
XM_017029451.1 Intron XP_016884940.1

View Full Product Details